1-Methylazetidine-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
068559-250MG
068559-500MG
101936-676EA
495.38
USD
101936-676
101936-678
1-Methylazetidine-3-carboxylic acid ≥95%
1-Methylazetidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₅H₉NO₂ MW: 115.1300 g/mol |
MDL Number:
MFCD11520621 CAS Number: 875629-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...