Azetidine-3-carbonitrile hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
azetidine-3-carbonitrile hydrochloride
068557-1G
068557-5G
101936-668EA
103.08
USD
101936-668
101936-670
Azetidine-3-carbonitrile hydrochloride ≥95%
Azetidine-3-carbonitrile hydrochloride
Hazard: Irritant
Formula:
C₄H₇ClN₂ MW: 118.57 g/mol |
MDL Number:
MFCD09749828 CAS Number: 345954-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...