3-(Methoxymethyl)azetidine trifluoroacetate ≥95%

Supplier: Matrix Scientific

068556-500MG 068556-1G 068556-5G
101936-662EA 500 USD
101936-662 101936-664 101936-666
3-(Methoxymethyl)azetidine trifluoroacetate ≥95%
3-(Methoxymethyl)azetidine trifluoroacetate

Hazard: Irritant

Formula: C₇H₁₂F₃NO₃
MDL Number: MFCD17016141
CAS Number: 1228230-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR