2,4-Dichloro-6-methylthieno[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

068555-250MG 068555-500MG
101936-658EA 253.85 USD
101936-658 101936-660
2,4-Dichloro-6-methylthieno[3,2-d]pyrimidine ≥95%
2,4-Dichloro-6-methylthieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₇H₄Cl₂N₂S
MDL Number: MFCD10000842
CAS Number: 35265-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR