Methyl-1-boc-3-azetidineacetate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(2-methoxy-2-oxoethyl)azetidine-1-carboxylate

068554-250MG 068554-500MG 068554-1G
101936-652EA 21.54 USD
101936-652 101936-654 101936-656
Methyl-1-boc-3-azetidineacetate ≥95%
Methyl-1-boc-3-azetidineacetate

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD11559001
CAS Number: 497160-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR