2-(1-Benzyloxycarbonylazetidin-3-yl)acetic acid ≥95%

Supplier: Matrix Scientific

068553-250MG 068553-500MG
101936-648EA 524.62 USD
101936-648 101936-650
2-(1-Benzyloxycarbonylazetidin-3-yl)acetic acid ≥95%
2-(1-Benzyloxycarbonylazetidin-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD02179029
CAS Number: 319470-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR