tert-Butyl-4-(chloromethyl)imidazole-1-carboxylate ≥95%

Supplier: Matrix Scientific

068551-1G 068551-5G
101936-640EA 692.31 USD
101936-640 101936-642
tert-Butyl-4-(chloromethyl)imidazole-1-carboxylate ≥95%
tert-Butyl-4-(chloromethyl)imidazole-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₂
MDL Number: MFCD08275019
CAS Number: 500782-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR