(1-Benzhydrylazetidin-3-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: [1-(diphenylmethyl)azetidin-3-yl]methanol

068550-1G 068550-5G
101936-636EA 113.85 USD
101936-636 101936-638
(1-Benzhydrylazetidin-3-yl)methanol ≥95%
(1-Benzhydrylazetidin-3-yl)methanol

Hazard: Irritant

Formula: C₁₇H₁₉NO
MW: 253.35 g/mol
MDL Number: MFCD03407522
CAS Number: 72351-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR