1-Benzhydrylazetidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Diphenylmethyl)azetidine-3-carboxylic Acid

068549-1G 068549-5G
101936-632EA 30.77 USD
101936-632 101936-634
1-Benzhydrylazetidine-3-carboxylic acid ≥95%
1-Benzhydrylazetidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₇NO₂
MW: 267.33 g/mol
MDL Number: MFCD01566538
CAS Number: 36476-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR