1-Benzhydrylazetidin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-1-(diphenylmethyl)azetidine

068547-1G 068547-5G
101936-624EA 3515.38 USD
101936-624 101936-626
1-Benzhydrylazetidin-3-amine ≥95%
1-Benzhydrylazetidin-3-amine

Hazard: Irritant

Formula: C₁₆H₁₈N₂
MW: 238.34 g/mol
MDL Number: MFCD03093386
CAS Number: 40432-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR