1-Azetidinecarboxylic acid-3-aminophenylmethyl ester ≥95%

Supplier: Matrix Scientific

068546-250MG 068546-500MG
101936-620EA 5163.08 USD
101936-620 101936-622
1-Azetidinecarboxylic acid-3-aminophenylmethyl ester ≥95%
1-Azetidinecarboxylic acid-3-aminophenylmethyl ester

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD07772068
CAS Number: 112257-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR