1-Azetidinecarboxylic acid-3-iodophenylmethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl 3-iodoazetidine-1-carboxylate, 1-Cbz-3-iodoazetidine

068545-250MG 068545-500MG
101936-616EA 349.23 USD
101936-616 101936-618
1-Azetidinecarboxylic acid-3-iodophenylmethyl ester ≥95%
1-Azetidinecarboxylic acid-3-iodophenylmethyl ester

Hazard: Irritant

Formula: C₁₁H₁₂INO₂
MW: 317.13 g/mol
MDL Number: MFCD09026510
CAS Number: 939759-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR