4-Pyrimidinecarboxylic acid, methyl ester ≥95%
Supplier: Matrix Scientific
068537-1G
068537-5G
101936-580EA
353.85
USD
101936-580
101936-582
4-Pyrimidinecarboxylic acid, methyl ester ≥95%
4-Pyrimidinecarboxylic acid, methyl ester
Hazard: Irritant
Formula:
C₆H₆N₂O₂ |
MDL Number:
MFCD00234184 CAS Number: 2450-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...