(6-Methylpyridazin-3-yl)methanamine ≥95%

Supplier: Matrix Scientific

068536-500MG 068536-1G
101936-576EA 2501.54 USD
101936-576 101936-578
(6-Methylpyridazin-3-yl)methanamine ≥95%
(6-Methylpyridazin-3-yl)methanamine

Hazard: Irritant

Formula: C₆H₉N₃
MDL Number: MFCD17015869
CAS Number: 1004972-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR