3-(Chloromethyl)pyridazine ≥95%

Supplier: Matrix Scientific

068534-250MG 068534-500MG 068534-1G
101936-566EA 784.62 USD
101936-566 101936-568 101936-570
3-(Chloromethyl)pyridazine ≥95%
3-(Chloromethyl)pyridazine

Hazard: Irritant

Formula: C₅H₅ClN₂
MDL Number: MFCD13189576
CAS Number: 41227-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR