2-(Chloromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

068531-1G 068531-5G
101936-554EA 230.77 USD
101936-554 102551-708
2-(Chloromethyl)pyrimidine ≥95%
2-(Chloromethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₅ClN₂
MDL Number: MFCD06796203
CAS Number: 54198-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR