2-(Chloromethyl)pyrazine ≥95%

Supplier: Matrix Scientific

068528-250MG 068528-500MG 068528-1G
101936-536EA 461.54 USD
101936-536 101936-538 101936-540
2-(Chloromethyl)pyrazine ≥95%
2-(Chloromethyl)pyrazine

Hazard: Irritant

Formula: C₅H₅ClN₂
MDL Number: MFCD10697790
CAS Number: 39204-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR