Pyrazin-2-ylmethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Pyrazinemethanol, 2-(Hydroxymethyl)pyrazine
068527-1G
068527-5G
101936-532EA
30.77
USD
101936-532
101936-534
Pyrazin-2-ylmethanol ≥95%
Pyrazin-2-ylmethanol
Hazard: Irritant
Formula:
C5H6N2O MW: 110.11 g/mol Melting Pt: 32…36 °C |
MDL Number:
MFCD01693938 CAS Number: 6705-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...