5-Pyrimidinecarbonitrile ≥95%

Supplier: Matrix Scientific

068524-1G 068524-5G
101936-520EA 146.15 USD
101936-520 101936-522
5-Pyrimidinecarbonitrile ≥95%
5-Pyrimidinecarbonitrile

Hazard: Irritant

Formula: C₅H₃N₃
MW: 105.1 g/mol
MDL Number: MFCD00235052
CAS Number: 40805-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR