5-Bromo-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

068520-250MG 068520-500MG
101936-500EA 872.31 USD
101936-500 101936-502
5-Bromo-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid ≥95%
5-Bromo-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD11040726
CAS Number: 916325-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR