6-Chloro-1H-indazole ≥95%

Supplier: Matrix Scientific

068519-1G 068519-5G
101936-496EA 104.62 USD
101936-496 101936-498
6-Chloro-1H-indazole ≥95%
6-Chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
MDL Number: MFCD07774234
CAS Number: 698-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR