1-Benzyl-2-methylpiperidin-4-amine ≥95%

Supplier: Matrix Scientific

068509-250MG 068509-500MG
101936-454EA 872.31 USD
101936-454 101936-456
1-Benzyl-2-methylpiperidin-4-amine ≥95%
1-Benzyl-2-methylpiperidin-4-amine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD14692665
CAS Number: 321345-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR