1-Benzyl-2-methylpiperidin-4-one ≥95%
Supplier: Matrix Scientific
068508-250MG
068508-500MG
101936-450EA
524.62
USD
101936-450
101936-452
1-Benzyl-2-methylpiperidin-4-one ≥95%
1-Benzyl-2-methylpiperidin-4-one
Hazard: Irritant
Formula:
C₁₃H₁₇NO |
MDL Number:
MFCD00480711 CAS Number: 203661-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...