(1S,5R)-8-Methyl-8-azabicyclo[3.2.1]octan-3-amine ≥95%

Supplier: Matrix Scientific

068506-1G 068506-5G
101936-440EA 169.23 USD
101936-440 101936-442
(1S,5R)-8-Methyl-8-azabicyclo[3.2.1]octan-3-amine ≥95%
(1S,5R)-8-Methyl-8-azabicyclo[3.2.1]octan-3-amine

Hazard: Irritant

Formula: C₈H₁₆N₂
MDL Number: MFCD00215843
CAS Number: 87571-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR