N-Benzyltropinone ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one, (1R,5S)-8-Benzyl-8-azabicyclo[3.2.1]octan-3-one

068505-1G 068505-5G 068505-25G
101936-434EA 175.38 USD
101936-434 101936-436 101936-438
N-Benzyltropinone ≥95%
N-Benzyltropinone

Hazard: Irritant

Formula: C₁₄H₁₇NO
MW: 215.30 g/mol
MDL Number: MFCD00151640
CAS Number: 28957-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR