(1S,5R)-3-Benzyloxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

068503-250MG 068503-500MG 068503-1G
101936-424EA 872.31 USD
101936-424 101936-426 101936-428
(1S,5R)-3-Benzyloxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid ≥95%
(1S,5R)-3-Benzyloxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₅NO₄
MDL Number: MFCD17214345
CAS Number: 134575-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR