Tetrahydropyran-4-acetic acid ≥95%
Supplier: Matrix Scientific
068502-1G
068502-5G
101936-420EA
30.77
USD
101936-420
101936-422
Tetrahydropyran-4-acetic acid ≥95%
Tetrahydropyran-4-acetic acid
Hazard: Irritant
Formula:
C₇H₁₂O₃ |
MDL Number:
MFCD01631204 CAS Number: 85064-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...