tert-Butyl-3-amino-3-methylazetidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3-amino-3-methylazetidine-1-carboxylate
068499-250MG
068499-500MG
068499-1G
101936-406EA
803.08
USD
101936-406
101936-408
101936-410
tert-Butyl-3-amino-3-methylazetidine-1-carboxylate ≥95%
tert-Butyl-3-amino-3-methylazetidine-1-carboxylate
Hazard: Irritant
Formula:
C₉H₁₈N₂O₂ MW: 186.25 g/mol |
MDL Number:
MFCD12408556 CAS Number: 1158758-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...