1-Methyl-1,4-diazepane ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methylhomopiperazine
068498-1G
068498-5G
101936-402EA
116.92
USD
101936-402
101936-404
1-Methyl-1,4-diazepane ≥95%
1-Methyl-1,4-diazepane
Hazard: Irritant
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol |
MDL Number:
MFCD00059810 CAS Number: 4318-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...