6-(Aminomethyl)-1H-pyrimidin-4-one dihydrochloride ≥95%

Supplier: Matrix Scientific

068495-250MG 068495-500MG 068495-1G
101936-394EA 1106.15 USD
101936-394 101936-396 101936-398
6-(Aminomethyl)-1H-pyrimidin-4-one dihydrochloride ≥95%
6-(Aminomethyl)-1H-pyrimidin-4-one dihydrochloride

Hazard: Irritant

Formula: C₅H₉Cl₂N₃O
MDL Number: MFCD19982769
CAS Number: 1269054-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR