2-(2,4-Dimethylphenyl)-1,2-dihydropyrazol-3-one ≥95%

Supplier: Matrix Scientific

068494-500MG 068494-1G
101936-390EA 1280 USD
101936-390 101936-392
2-(2,4-Dimethylphenyl)-1,2-dihydropyrazol-3-one ≥95%
2-(2,4-Dimethylphenyl)-1,2-dihydropyrazol-3-one

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD19443944
CAS Number: 1245204-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR