tert-Butyl-10-oxo-4,9-diazaspiro[4.5]decane-4-carboxylate ≥95%

Supplier: Matrix Scientific

068492-250MG 068492-500MG
101936-382EA 524.62 USD
101936-382 101936-384
tert-Butyl-10-oxo-4,9-diazaspiro[4.5]decane-4-carboxylate ≥95%
tert-Butyl-10-oxo-4,9-diazaspiro[4.5]decane-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₂N₂O₃
MDL Number: MFCD13184421
CAS Number: 886449-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR