1H-Pyrrolo[2,3-b]pyridine-3-propanoic acid ≥95%

Supplier: Matrix Scientific

068488-1G 068488-5G
101936-366EA 1107.69 USD
101936-366 101936-368
1H-Pyrrolo[2,3-b]pyridine-3-propanoic acid ≥95%
1H-Pyrrolo[2,3-b]pyridine-3-propanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD09756478
CAS Number: 27663-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR