1-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine ≥95%

Supplier: Matrix Scientific

068487-250MG 068487-500MG 068487-1G
101936-360EA 263.08 USD
101936-360 101936-362 101936-364
1-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine ≥95%
1-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine

Hazard: Irritant

Formula: C₁₈H₂₆BNO₂
MDL Number: MFCD11840350
CAS Number: 1313738-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR