Phenyl(1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridin-2-yl)methanon ≥95%

Supplier: Matrix Scientific

068485-500MG 068485-1G
101936-356EA 1773.85 USD
101936-356 101936-358
Phenyl(1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridin-2-yl)methanon ≥95%
Phenyl(1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridin-2-yl)methanon

Hazard: Irritant

Formula: C₂₀H₁₄N₂O₃S
MDL Number: MFCD12912595
CAS Number: 1198283-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR