Phenyl(1H-pyrrolo[2,3-b]pyridin-2-yl)methanone ≥95%

Supplier: Matrix Scientific

068484-500MG 068484-1G
101936-352EA 1687.69 USD
101936-352 101936-354
Phenyl(1H-pyrrolo[2,3-b]pyridin-2-yl)methanone ≥95%
Phenyl(1H-pyrrolo[2,3-b]pyridin-2-yl)methanone

Hazard: Irritant

Formula: C₁₄H₁₀N₂O
MDL Number: MFCD12912596
CAS Number: 1198284-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR