cis-tert-Butyl-N-(3-aminocyclobutyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: cis-tert-Butyl 3-aminocyclobutanecarbamate

068481-250MG 068481-500MG 068481-1G
101936-340EA 1064.33 USD
101936-340 101936-342 101936-344
cis-tert-Butyl-N-(3-aminocyclobutyl)carbamate ≥95%
cis-tert-Butyl-N-(3-aminocyclobutyl)carbamate

Hazard: Irritant

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
MDL Number: MFCD11099896
CAS Number: 1212395-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR