3-Aminocyclobutanone hydrochloride ≥95%

Supplier: Matrix Scientific

068480-250MG 068480-1G
101936-334EA 306.15 USD
101936-334 101936-338
3-Aminocyclobutanone hydrochloride ≥95%
3-Aminocyclobutanone hydrochloride

Hazard: Irritant

Formula: C₄H₈ClNO
MDL Number: MFCD12923220
CAS Number: 1035374-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR