tert-Butyl-6-formyl-3,4-dihydro-1H-pyrrolo[1,2-a]pyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

068478-250MG 068478-500MG 068478-1G
101936-324EA 466.15 USD
101936-324 101936-326 101936-328
tert-Butyl-6-formyl-3,4-dihydro-1H-pyrrolo[1,2-a]pyrazine-2-carboxylate ≥95%
tert-Butyl-6-formyl-3,4-dihydro-1H-pyrrolo[1,2-a]pyrazine-2-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MW: 250.3 g/mol
MDL Number: MFCD13195336
CAS Number: 1174068-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR