3-Cyano-1H-indole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

068477-500MG 068477-1G
101936-320EA 1426.15 USD
101936-320 101936-322
3-Cyano-1H-indole-5-carboxylic acid ≥95%
3-Cyano-1H-indole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆N₂O₂
MDL Number: MFCD07776627
CAS Number: 889942-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR