7,8-Dihydro-5(6H)-quinolinone ≥95%

Supplier: Matrix Scientific

Synonyms: 7,8-Dihydroquinolin-5(6H)-one

068476-500MG 068476-1G
101936-316EA 369.21 USD
101936-316 101936-318
7,8-Dihydro-5(6H)-quinolinone ≥95%
7,8-Dihydro-5(6H)-quinolinone

Hazard: Irritant

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD02181161
CAS Number: 53400-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR