4-(4-Ethyl-1-piperazinyl)-3-fluoro-benzenamine ≥95%

Supplier: Matrix Scientific

068474-500MG 068474-1G
101936-312EA 233.85 USD
101936-312 101936-314
4-(4-Ethyl-1-piperazinyl)-3-fluoro-benzenamine ≥95%
4-(4-Ethyl-1-piperazinyl)-3-fluoro-benzenamine

Hazard: Irritant

Formula: C₁₂H₁₈FN₃
MDL Number: MFCD08699561
CAS Number: 873537-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR