2H-2,7-Naphthyridin-1-one ≥95%
Supplier: Matrix Scientific
068473-250MG
068473-500MG
068473-1G
101936-306EA
315.38
USD
101936-306
101936-308
101936-310
2H-2,7-Naphthyridin-1-one ≥95%
2H-2,7-Naphthyridin-1-one
Hazard: Irritant
Formula:
C₈H₆N₂O |
MDL Number:
MFCD11877928 CAS Number: 67988-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...