1,3-Dihydropyrrolo[2,3-c]pyridin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Dihydro-2H-pyrrolo[2,3-c]pyridin-2-one

068472-1G 068472-5G
101936-302EA 1500 USD
101936-302 101936-304
1,3-Dihydropyrrolo[2,3-c]pyridin-2-one ≥95%
1,3-Dihydropyrrolo[2,3-c]pyridin-2-one

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD08234948
CAS Number: 54415-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR