1,3-Dihydropyrrolo[3,2-b]pyridin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1H-Pyrrolo[3,2-b]pyridin-2(3H)-one

068471-250MG 068471-500MG
101936-298EA 553.85 USD
101936-298 101936-300
1,3-Dihydropyrrolo[3,2-b]pyridin-2-one ≥95%
1,3-Dihydropyrrolo[3,2-b]pyridin-2-one

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD02179615
CAS Number: 32501-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR