1H-Indazole-6-carbonitrile ≥95%

Supplier: Matrix Scientific

068470-1G 068470-5G
101936-294EA 1076.92 USD
101936-294 101936-296
1H-Indazole-6-carbonitrile ≥95%
1H-Indazole-6-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MDL Number: MFCD04973398
CAS Number: 141290-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR