6-Bromo-1-methyl-1H-indazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

068468-250MG 068468-500MG 068468-1G
101936-282EA 1076.92 USD
101936-282 101936-284 101936-286
6-Bromo-1-methyl-1H-indazole-3-carboxylic acid ≥95%
6-Bromo-1-methyl-1H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MDL Number: MFCD15071436
CAS Number: 1021859-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR