Methyl-5-bromo-1H-indazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-bromo-1H-indazole-3-carboxylate
068467-1G
068467-5G
101936-278EA
815.38
USD
101936-278
101936-280
Methyl-5-bromo-1H-indazole-3-carboxylate ≥95%
Methyl 5-bromo-1H-indazole-3-carboxylate
Hazard: Irritant
Formula:
C₉H₇BrN₂O₂ |
MDL Number:
MFCD07371572 CAS Number: 78155-74-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...