Methyl-3-bromo-1H-indazole-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-bromo-1H-indazole-6-carboxylate

068465-1G 068465-5G
101936-268EA 230.77 USD
101936-268 101936-270
Methyl-3-bromo-1H-indazole-6-carboxylate ≥95%
Methyl 3-bromo-1H-indazole-6-carboxylate

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.0700 g/mol
MDL Number: MFCD11499017
CAS Number: 192945-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR