1-Methylindazole-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-1H-indazole-5-carboxylic acid
068464-250MG
068464-500MG
068464-1G
101936-262EA
321.69
USD
101936-262
101936-264
101936-266
1-Methylindazole-5-carboxylic acid ≥95%
1-Methylindazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₉H₈N₂O₂ MW: 176.18 g/mol |
MDL Number:
MFCD11901714 CAS Number: 1176754-31-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...